missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey Anti-Mouse IgG Biotinylated Antibody, R&D Systems™

Description
IgG Polyclonal specifically detects IgG in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | IgG |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Biotin |
| Dilution | Western Blot |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Gene Symbols | IGHG1 |
| Host Species | Donkey |
| Immunogen | Mouse IgG |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?