missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, DyLight 488 (Pre-adsorbed), Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Fluorophore-linked immunosorbent assay.
Specifications
Specifications
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Dilution | Western Blot 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:5000, Immunohistochemistry-Frozen, Dot Blot, Fluorophore-linked immunosorbent assay 1:20000 |
| Purification Method | Affinity Purified |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?