missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, DyLight 405 (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-72853
185.30 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| DyLight 405 | |
| Donkey | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Reconstitute with 100 ul deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:5000, Fluorophore-linked immunosorbent assay 1:20000 | |
| Goat IgG whole molecule | |
| 0.1 mg | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Chicken, Guinea Pig, Hamster, Horse, Mouse, Rabbit, and Rat Serum Proteins. No reaction was observed against Chicken, Guinea Pig, Hamster, Horse, Mouse, Rabbit and Rat Serum Proteins. This antibody will react with heavy chains of Goat IgG and with light chains of most Goat immunoglobulins. | |
| Goat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction