missing translation for 'onlineSavingsMsg'
Learn More

Donkey anti-Goat IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™

Product Code. 18225333 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18225333 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18225333 Supplier Novus Biologicals Supplier No. NBP174812
  • ONLINE EXCLUSIVE PRICE
    286.00€
    269.85€ / 1mg
    Save 16.15€
    5% Off
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.50mg/mL
Conjugate Alkaline Phosphatase (AP)
Dilution Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin
Host Species Donkey
Immunogen Purified Goat IgG (H&L)
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Based on IEP, this antibody reacts with heavy gamma chains on goat IgG and light chains on all goat immunoglobulins
Target Species Goat
Content And Storage Store at 4C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20C. Avoid repeated freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.