missing translation for 'onlineSavingsMsg'
Learn More

Donkey Anti-Goat IgG H&L (Cy5 ™) preadsorbed, Abcam

Product Code. 32116653
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116653 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116653 Supplier Abcam Supplier No. ab6566500ug
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Donkey Polyclonal Antibody

Suitable for WB, Flow Cyt, ICC, IHC-Fr, IHC-P, ICC/IF, ELISA. Preadsorbed to minimise non-specific binding and high background staining. Cited in 44 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifikationer

Antigen Goat IgG
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE), Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Cyanine5
Formulation pH: 6.5, Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Tripotassium orthophosphate
Host Species Donkey
Immunogen Goat IgG
Purification Method Affinity purification
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage 4°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Molecule Goat IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.