missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€ - 593.00€
Specifications
| Antigen | DOK1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18292321
|
Novus Biologicals
NBP2-55613 |
100 μL |
593.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681888
|
Novus Biologicals
NBP2-55613-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DOK1 Polyclonal specifically detects DOK1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| DOK1 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 1796 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AQGNDIFQAVETAIHRQKAQGKAGQGHDVLRADSHEGEVAEGKLPSPPGPQELLDSPPALYAEPLDSLRIAPCPSQDSLYSDPLDSTSAQAGEGVQRKKPLYWDLYEHAQQQLLKAKLTDPKEDPIYDEPEG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| docking protein 1, docking protein 1 (downstream of tyrosine kinase 1), docking protein 1, 62kD (downstream of tyrosine kinase 1), docking protein 1, 62kDa (downstream of tyrosine kinase 1), Downstream of tyrosine kinase 1, MGC117395, MGC138860, p62(dok), P62DOK, pp62 | |
| DOK1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title