missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK8 Monoclonal Antibody (OTI16G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801795
This item is not returnable.
View return policy
Description
The Dedicator of cytokinesis protein 8 (DOCK8) is a member of the DOCK180 family of guanine nucleotide exchange factors. DOCK8 plays an essential role in humoral immune responses and is important in the proper formation of the B cell immunological synapse. Mutations in this gene result in the autosomal recessive form of the hyper-IgE syndrome.Specifications
| DOCK8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NF50 | |
| DOCK8 | |
| Human recombit protein fragment corresponding to amino acids 84-405 of human DOCK8 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI16G2 | |
| Unconjugated | |
| DOCK8 | |
| HEL-205, MRD2, ZIR8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 81704 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction