Learn More
Thermo Scientific™ dNTP Mixes Promotion
Product Code. p-7068036 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.2mL (Total: 8μmol)
5 x 1mL (Total: 200μmol)
Unit Size:
0.2mL
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15777536 0.2mL (Total: 8μmol) 0.2mL
15797536 5 x 1mL (Total: 200μmol) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15777536 Supplier Thermo Scientific™ Supplier No. R0191
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reduce the number of pipetting steps and the risk of reaction setup errors with these dNTP mixes, premixed aqueous solutions of dATP, dTTP, dCTP, and dGTP.
dNTP Mixes contain premixed aqueous solutions of dATP, dCTP, dGTP and dTTP, each at a final concentration of 2, 10 or 25 mM. The nucleotides have greater than 99% purity, are free of nuclease activities, human and E. coli DNA. Mixes offer the possibility to reduce the number of pipetting steps and the risk of reaction set up errors. They are designed for many different molecular biology applications.
Highlights
  • Greater than 99% purity confirmed by HPLC
  • Free of human and E. coli DNA
  • Highly stable
    • stable for years at -20°C
    • Stable after multiple freeze-thaw cycles
    • Up to 95% of dNTPs remain in triphosphate form even after 7 weeks at room temperature
    • Up to 90% of dNTPs remain in triphosphate form after 30 cycles of PCR (1 minute at 94°C; 3 minutes at 72°C)
  • Application tested in:
    • Long range PCR (40 kb)
    • cDNA synthesis and RT-PCR
    • Real-time PCR
    • Standard PCR
    • High fidelity PCR
TRUSTED_SUSTAINABILITY
Specifications
Purity or Quality Grade >99% (by HPLC)
Quantity 0.2mL (Total: 8μmol)
For Use With (Application) Real Time PCR (qPCR), RT-PCR, Standard PCR