missing translation for 'onlineSavingsMsg'
Learn More

DNMT3B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. 16895285 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16895285 150 μL 150µL
16805295 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16895285 Supplier Proteintech Supplier No. 269711AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The DNMT3B antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human DNMT3B. This antibody recognizes human, mouse antigen. The DNMT3B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.

Specifications

Antigen DNMT3B
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Concentration 467 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 1789
Target Species Human, Mouse
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.