missing translation for 'onlineSavingsMsg'
Learn More

DNMT1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-392R

Product Code. 18728612

  • 633.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DNMT1 Polyclonal antibody specifically detects DNMT1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNMT1
Polyclonal
Western Blot
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human DNA-Methyltranserase 1 using the numbering given in entry NP_001370.1 (GeneID 1786).
0.1 mL
Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
1786
Store at 4°C in the dark.
IgG
Western Blot
DyLight 550
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.