missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNM1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68009-1-IG-150UL
This item is not returnable.
View return policy
Description
The DNM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DNM1. This antibody recognizes human, rat, mouse, rabbit, pig, chicken antigen. The DNM1 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| DNM1 | |
| Monoclonal | |
| 1000 μg/mL | |
| DNM | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Rat, Mouse, Rabbit, Pig, Chicken | |
| 100 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1B2A11 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 1759 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction