missing translation for 'onlineSavingsMsg'
Learn More

DNASE1L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16801984 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16801984 20 μL 20µL
16891974 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16801984 Supplier Proteintech Supplier No. 136531AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a deoxyribonuclease protein that shows high sequence similarity to DNase I. The encoded protein is localized to the endoplasmic reticulum and modified by N-linked glycosylation. Alternate transcriptional splice variants encoding the same protein have been observed.

Specifications

Antigen DNASE1L1
Applications Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DNASE1L1
Gene Accession No. P49184, Q2QDE7, Q9D7J6
Gene Alias Deoxyribonuclease 1 like 1, deoxyribonuclease I like 1, DNAS1L1, DNase I like 1, DNase X, DNASE1L1, DNASEX, DNL1L, Muscle specific DNase I like, XIB
Gene Symbols DNASE1L1
Host Species Rabbit
Immunogen DNASE1L1 Fusion Protein Ag4222
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1774, 363522, 69537
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.