missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92922-0.02ml
This item is not returnable.
View return policy
Description
DNAL1 Polyclonal antibody specifically detects DNAL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| DNAL1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:100 | |
| C14orf168MGC12435,1700010H15RiK, chromosome 14 open reading frame 168, dynein light chain 1, axonemal, dynein, axonemal, light chain 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 41-190 of human DNAL1 (NP_113615.2). DASLSMLANCEKLSLSTNCIEKIANLNGLKNLRILSLGRNNIKNLNGLEAVGDTLEELWISYNFIEKLKGIHIMKKLKILYMSNNLVKDWAEFVKLAELPCLEDLVFVGNPLEEKHSAENNWIEEATKRVPKLKKLDGTPVIKGDEEEDN | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Neurodegeneration, Neuroscience, Signal Transduction | |
| 83544 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction