Learn More
DNAJB5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16453-1-AP-150UL
This item is not returnable.
View return policy
Description
DNAJB5 belongs to the evolutionarily conserved DNAJ/HSP40 protein family. For background information on the DNAJ family.Specifications
| DNAJB5 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB5 | |
| DNAJB5, Heat shock protein cognate 40, Heat shock protein Hsp40 2, Heat shock protein Hsp40 3, Hsc40, KIAA1045 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25822, 313811, 56323 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75953, O89114 | |
| Dnajb5 | |
| DNAJB5 Fusion Protein Ag9341 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |