missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13064-1-AP-150UL
This item is not returnable.
View return policy
Description
DnaJ heat shock induced proteins are from the bacterium Escherichia coli and are under the control of the htpR regulatory protein. The DnaJ proteins play a critical role in the HSP 70 chaperone machine by interacting with HSP 70 to stimulate ATP hydrolysis. The proteins contain cysteine rich regions that are composed of zinc fingers that form a peptide binding domain responsible for the chaperone function. DnaJ proteins are important mediators of proteolysis and are involved in the regulation of protein degradation, exocytosis and endocytosis. DnaJB4 (DnaJ homolog subfamily B member 4), also known as HLJ1, is expressed in skeletal muscle, heart and pancreas, and lower expression in brain, placenta and liver.Specifications
| DNAJB4 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB4 | |
| DjB4, DNAJB4, DNAJW, Heat shock protein 40 homolog, HLJ1, HSP40 homolog, Human liver DnaJ like protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11080, 295549, 67035 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9D832, Q9UDY4 | |
| Dnajb4 | |
| DNAJB4 Fusion Protein Ag3756 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction