missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL1 Antibody (306), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89936CL1
This item is not returnable.
View return policy
Description
DLL1 Monoclonal antibody specifically detects DLL1 in Human samples. It is validated for ELISA
Specifications
| DLL1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human DLL1 (Accession#: NP_005609.3; Met 1-Gly 540). | |
| 0.1 mL | |
| Neuroscience, Signal Transduction, Stem Cell Signaling Pathway | |
| 28514 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 306 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction