missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35894-20ul
This item is not returnable.
View return policy
Description
DLC1 Polyclonal antibody specifically detects DLC1 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DLC1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| ARHGAP7 STARD12 deleted in liver cancer 1 variant 2, deleted in liver cancer 1, Deleted in liver cancer 1 protein, DLC-1, FLJ21120, HP, HP protein, KIAA1723, p122-RhoGAP, rho GTPase-activating protein 7, Rho-GTPase-activating protein 7, Rho-type GTPase-activating protein 7, StARD12, StAR-related lipid transfer (START) domain containing 12, StAR-related lipid transfer protein 12, START domain-containing protein 12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 204-463 of human DLC1 (NP_079043.3).,, Sequence:, DAPKVNAVDTLNVKDIAPEKQLLNSAVIAQQRRKPDPPKDENERSTCNVVQNEFLDTPCTNRGLPLLKTDFGSCLLQPPSCPNGMSAENGLEKSGFSQHQNKSPPKVKAEDGMQCLQLKETLATQEPTDNQVRLRKRKEIREDRDRARLDSMVLLIMKLDQLDQDIENALSTSSSPSGTPTNLRRHVPDLESGSESGADTISVNQTRVNLSSDTESTDLPSSTPVANSGTKPKTTAIQGISEKEKAGKLTFWFCFLANLF | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10395 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction