missing translation for 'onlineSavingsMsg'
Learn More

DjC9 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18610842 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18610842 0.02 mL 0.02mL
18612692 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18610842 Supplier Novus Biologicals Supplier No. NBP2927140.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DjC9 Polyclonal antibody specifically detects DjC9 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DjC9
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 9, dnaJ homolog subfamily C member 9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DNAJC9 (NP_056005.1). MGLLDLCEEVFGTADLYRVLGVRREASDGEVRRGYHKVSLQVHPDRVGEGDKEDATRRFQILGKVYSVLSDREQRAVYDEQGTVDEDSPVLTQDRDWEAYWRLLFKKISLEDIQAFEKTYKGSEEELADIKQAYLDFKGDMDQIMESVLCVQYTEEPRIRNIIQQAIDAGEVPSYNAFVKESKQKMNARKRRAQEEAKEAEMSRKELGLDEGVDSLKAAIQSRQKDRQKEMDNFLAQMEAKYCKSSKGGGKKSALKKEKK
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23234
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.