Learn More
Distemper Virus Antibody (5-4), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB110-2604AF350
Product Code. 18789652
  • 519.00€ / 0.20mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
Distemper Virus Monoclonal antibody specifically detects Distemper Virus in Virus samples. It is validated for ELISA, Immunoassay, ELISA, Lateral Flow
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Distemper Virus
Monoclonal
Alexa Fluor 350
50mM Sodium Borate
Mouse
Protein A purified
RUO
Virus
Purified
ELISA, Immunoassay, ELISA, Lateral Flow
2024-05-04 00:00:00
ELISA, Immunoassay, Sandwich ELISA, Lateral Flow Assay
CDV, Canine Distemper Virus
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Canine distemper virus (CDV).
0.2 mL
Primary
Store at 4C in the dark.
IgG2a