Learn More
Distemper Virus Antibody (5-4), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-2604AF350
This item is not returnable.
View return policy
Description
Distemper Virus Monoclonal antibody specifically detects Distemper Virus in Virus samples. It is validated for ELISA, Immunoassay, ELISA, Lateral Flow
Specifications
| Distemper Virus | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Immunoassay, ELISA, Lateral Flow | |
| 2024-05-04 00:00:00 | |
| ELISA, Immunoassay, Sandwich ELISA, Lateral Flow Assay | |
| CDV, Canine Distemper Virus | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Canine distemper virus (CDV). | |
| 0.2 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |