missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DISP2 Monoclonal Antibody (758204)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561953
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
DISP2, also known as dispatched homolog 2, is a protein that plays a critical role in the regulation of the hedgehog signaling pathway. It is involved in the transport and release of hedgehog ligands, which are important for cellular differentiation and tissue development. Mutations in the DISP2 gene have been associated with various developmental disorders, including holoprosencephaly and cleft lip and palate. DISP2 is also being studied as a potential therapeutic target in cancer, as the hedgehog pathway is frequently dysregulated in various types of tumors.
Specifications
| DISP2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| A7MBM2 | |
| DISP2 | |
| E. coli-derived recombinant human DISP2, Arg192-Lys477. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 758204 | |
| Unconjugated | |
| DISP2 | |
| AI840597; B230210L08Rik; C15orf36; DISP2; dispatched B; dispatched homolog 2; dispatched homolog 2 (Drosophila); dispatched RND transporter family member 2; DISPB; HsT16908; KIAA1742; LINC00594; mKIAA1742; Protein dispatched homolog 2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 85455 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction