missing translation for 'onlineSavingsMsg'
Learn More

Disialosyl LEA Antigen Antibody (FH7), Novus Biologicals™

Product Code. 18136234 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18136234 0.1 mL 0.1mL
18102103 0.025 mL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18136234 Supplier Novus Biologicals Supplier No. NBP229808

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Disialosyl LEA Antigen Monoclonal specifically detects Disialosyl LEA Antigen in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Disialosyl LEA Antigen
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone FH7
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation Ascites with No Preservative
Gene Alias Disialosyl LEA, Disialosyl LEA antigen
Host Species Mouse
Immunogen Purified Disialosyl Lea antigen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Ascites
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.