missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Dipeptidyl Peptidase IV, MP BIOMEDICALS
Shop alle MP Biomedical produkter
Klik for at se tilgængelige muligheder
:
Dipeptidyl Peptidase IV
Beskrivelse
- Dipeptidyl Peptidase Iv (Dpp-Iv) Human
Application Notes:
- Dipeptidyl peptidase-4 plays an important role in tumor biology, and is useful as a marker for various cancers.
- It appears to work as a suppressor in the development of cancer and tumors.
Usage Statement:
- Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Key Applications:
- Apoptosis
Tekniske data
Tekniske data
| Form | Lyophilized Solid |
| Quantity | 10 MU |
| Synonym | CD26 |
| Format | Standard |
| Product Type | Protease |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion