missing translation for 'onlineSavingsMsg'
Learn More

Dinosam, TRC

Product Code. 30010975
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010975 50 mg 50mg
30138522 500 mg 500mg
30003257 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010975 Supplier Toronto Research Chemicals Supplier No. TRCD40973050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4097-36-3
Formula Weight 254.0903
InChI Formula InChI=1S/C11H14N2O5/c1-3-4-7(2)9-5-8(12(15)16)6-10(11(9)14)13(17)18/h5-7,14H,3-4H2,1-2H3
IUPAC Name 2,4-dinitro-6-pentan-2-ylphenol
Molecular Formula C11 H14 N2 O5
Molecular Weight (g/mol) 254.2393
Percent Purity >95%
SMILES CCCC(C)c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]
Synonym 2-(1-Methylbutyl)-4,6-dinitrophenol, 2,4-Dinitro-6-(1-methylbutyl)phenol, 2,4-Dinitro-6-(pentan-2-yl)phenol, 2-(1-Methyl-n-butyl)-4,6-dinitrophenol, 2-(2-Hydroxy-3,5-dinitrophenyl)pentane, 2-sec-Amyl-4,6-dinitrophenol, Chemox General, DNAP, DNOSAP, Dinosam, NSC 21493
Chemical Name or Material Dinosam
Grade HPLC
Show More Show Less

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.