missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimetindene Maleate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Dimetindene Maleate
Specifications
Specifications
| Chemical Name or Material | Dimetindene maleate |
| CAS | 3614-69-5 |
| Analyte or Component Names | Dimetindene maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 N2 . C4 H4 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Dimetindene maleate, N,N-Dimethyl-2-[3-[(RS)-1-(pyridin-2-yl)ethyl]-1H-inden-2-yl]ethanamine (Z)-butenedioate, 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-, (2Z)-2-butenedioate (1:1), 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-, (Z)-2-butenedioate (1:1), Pyridine, 2-[1-[2-(dimethylaminoethyl)inden-3-yl]ethyl]-, maleate (6CI), Pyridine, 2-[1-[2-[2-(dimethylamino)ethyl]inden-3-yl]ethyl]-, maleate (1:1) (7CI,8CI), 2-[1-[2-(2-Dimethylaminoethyl)inden-3-yl]ethyl]pyridine maleate, Dimethinden maleate, Dimethindene maleate, Dimethpyrindene maleate, Fenistil, Fenistil-retard, Fenostil, Forhistal maleate, NSC 107677, Su-6518 |
| InChI Formula | InChI=1S/C20H24N2.C4H4O4/c1-15(19-10-6-7-12-21-19)20-17(11-13-22(2)3)14-16-8-4-5-9-18(16)20;5-3(6)1-2-4(7)8/h4-10,12,15H,11,13-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CC(C1=C(CCN(C)C)Cc2ccccc12)c3ccccn3.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N,N-dimethyl-2-[3-(1-pyridin-2-ylethyl)-1H-inden-2-yl]ethanamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction