missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl-(tetrahydro-3,3-diphenyl-2-furilidine)ammonium Bromide, Mikromol™
Description
Dimethyl-(tetrahydro-3,3-diphenyl-2-furilidine)ammonium Bromide
Specifications
Specifications
| Chemical Name or Material | Dimethyl-(tetrahydro-3,3-diphenyl-2-furilidine)ammonium Bromide |
| CAS | 37743-18-3 |
| Analyte or Component Names | Dimethyl-(tetrahydro-3,3-diphenyl-2-furilidine)ammonium Bromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H20 N O . Br |
| Certifications/Compliance | ISO 17025 |
| Synonym | Dimethyl-(tetrahydro-3,3-diphenyl-2-furilidine)ammonium Bromide |
| InChI Formula | InChI=1S/C18H20NO.BrH/c1-19(2)17-18(13-14-20-17,15-9-5-3-6-10-15)16-11-7-4-8-12-16;/h3-12H,13-14H2,1-2H3;1H/q+1;/p-1 |
| SMILES | [Br-].C[N+](=C1OCCC1(c2ccccc2)c3ccccc3)C |
| IUPAC Name | (3,3-diphenyloxolan-2-ylidene)-dimethylazanium;bromide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?