missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diltiazem Sulphoxide, Mikromol
Description
Diltiazem Sulphoxide, Mikromol
Specifications
Specifications
| Chemical Name or Material | Diltiazem Sulphoxide |
| CAS | 370094-12-5 |
| Analyte or Component Names | Diltiazem Sulphoxide |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C22 H26 N2 O5 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Diltiazem Sulphoxide |
| InChI Formula | InChI=1S/C22H26N2O5S/c1-15(25)29-20-21(16-9-11-17(28-4)12-10-16)30(27)19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3/h5-12,20-21H,13-14H2,1-4H3/t20-,21+,30?/m1/s1 |
| Solution Type | Neat |
| SMILES | COc1ccc(cc1)[C@H]2[C@@H](OC(=O)C)C(=O)N(CCN(C)C)c3ccccc3S2=O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?