missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-4-methylthio-2-pentanone, 99%, Thermo Scientific™

Product Code. 11439557
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11439557

Brand: Thermo Scientific Alfa Aesar B22058.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23550-40-5
Molecular Formula C7H14OS
Molecular Weight (g/mol) 146.25
MDL Number MFCD00049253
InChI Key DHANVOSHQILVNQ-UHFFFAOYSA-N
Synonym 4-methylthio-4-methyl-2-pentanone, 4-methyl-4-methylthio pentan-2-one, 2-pentanone, 4-methyl-4-methylthio, 4-methyl-4-methylthio-2-pentanone, unii-g69hk72q3y, 4-methyl-4-methylsulfanyl pentan-2-one, cassis pentanone, fema no. 3376, corps cassis
PubChem CID 4682590
IUPAC Name 4-methyl-4-methylsulfanylpentan-2-one
SMILES CSC(C)(C)CC(C)=O

Specifications

Density 0.965
Boiling Point 80°C to 82°C (10 mmHg)
Flash Point 72°C (162°F)
Refractive Index 1.472
Quantity 25 g
Formula Weight 146.25
Percent Purity 99%
Chemical Name or Material 4-Methyl-4-methylthio-2-pentanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.