CAS RN 23550-40-5
CAS RN 23550-40-5
IUPAC Name:
4-methyl-4-(methylsulfanyl)pentan-2-one
Synonyms:
4-methyl-4-(methylsulfanyl)pentan-2-one
Molecular Weight (g/mol):
146.25
Molecular Formula:
C7H14OS
InChi Key:
DHANVOSHQILVNQ-UHFFFAOYSA-N
SMILES:
CSC(C)(C)CC(C)=O
1
–
1
of
1
results
1
4-Methyl-4-methylthio-2-pentanone, 99%, Thermo Scientific™
CAS: 23550-40-5 Molecular Formula: C7H14OS Molecular Weight (g/mol): 146.25 MDL Number: MFCD00049253 InChI Key: DHANVOSHQILVNQ-UHFFFAOYSA-N Synonym: 4-methylthio-4-methyl-2-pentanone,4-methyl-4-methylthio pentan-2-one,2-pentanone, 4-methyl-4-methylthio,4-methyl-4-methylthio-2-pentanone,unii-g69hk72q3y,4-methyl-4-methylsulfanyl pentan-2-one,cassis pentanone,fema no. 3376,corps cassis PubChem CID: 4682590 IUPAC Name: 4-methyl-4-methylsulfanylpentan-2-one SMILES: CSC(C)(C)CC(C)=O