missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Difloxacin Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Difloxacin Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Difloxacin hydrochloride |
| CAS | 91296-86-5 |
| Analyte or Component Names | Difloxacin hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H19 F2 N3 O3 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | 3-Quinolinecarboxylic acid, 6-fluoro-1-(4-fluorophenyl)-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-, hydrochloride (1:1), 3-Quinolinecarboxylic acid, 6-fluoro-1-(4-fluorophenyl)-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-, monohydrochloride (9CI), Difloxacin hydrochloride |
| InChI Formula | InChI=1S/C21H19F2N3O3.ClH/c1-24-6-8-25(9-7-24)19-11-18-15(10-17(19)23)20(27)16(21(28)29)12-26(18)14-4-2-13(22)3-5-14;/h2-5,10-12H,6-9H2,1H3,(H,28,29);1H |
| SMILES | Cl.CN1CCN(CC1)c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)c4ccc(F)cc4 |
| IUPAC Name | 6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction