missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4RS)-2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, Mikromol™

Product Code. 30049666
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049666 25 mg 25mg
30050560 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049666 Supplier Mikromol Supplier No. MM0383.100025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diethyl (4RS)-2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl (4RS)-2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
CAS 140171-65-9
Analyte or Component Names Diethyl (4RS)-2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Shipping Condition Room Temperature
Molecular Formula C21 H27 Cl N2 O5
Certifications/Compliance ISO 17025
Synonym Amlodipine Besilate Imp. E (EP), Amlodipine Imp. E (EP), Diethyl (4RS)-2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Related Compound E, Amlodipine Besilate Impurity E, Amlodipine Impurity E
InChI Formula InChI=1S/C21H27ClN2O5/c1-4-28-20(25)17-13(3)24-16(12-27-11-10-23)19(21(26)29-5-2)18(17)14-8-6-7-9-15(14)22/h6-9,18,24H,4-5,10-12,23H2,1-3H3
SMILES CCOC(=O)C1=C(C)NC(=C(C1c2ccccc2Cl)C(=O)OCC)COCCN
IUPAC Name diethyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 422.9
Quantity 25 mg
Formula Weight 422.1608 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.