missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Diethylamine (Diethylammonium Diclofenac), Mikromol™

Product Code. 18092890
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18092890 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18092890 Supplier Mikromol Supplier No. MM0006.25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diclofenac Diethylamine (Diethylammonium Diclofenac)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diclofenac Diethylamine
CAS 78213-16-8
Analyte or Component Names Diclofenac Diethylamine
Shipping Condition Room Temperature
Molecular Formula C14 H11 Cl2 N O2 . C4 H11 N
Certifications/Compliance ISO 17034
Synonym Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, compd. with N-ethylethanamine (1:1), Ethanamine, N-ethyl-, 2-[(2,6-dichlorophenyl)amino]benzeneacetate (9CI), Atomo Desinflamante, Diclofenac diethylamine, Diclofenac diethylamine salt, Diclofenac diethylammonium salt, Voltaren Emulgel, Voltaren Schmerzgel
InChI Formula InChI=1S/C14H11Cl2NO2.C4H11N/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;1-3-5-4-2/h1-7,17H,8H2,(H,18,19);5H,3-4H2,1-2H3
SMILES CCNCC.OC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name 2-[2-(2,6-dichloroanilino)phenyl]acetic acid;N-ethylethanamine
Molecular Weight (g/mol) 369.29
Quantity 250 mg
Formula Weight 368.1058 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.