missing translation for 'onlineSavingsMsg'
Learn More

DHX58 Antibody - BSA Free, Novus Biologicals™

Product Code. 18570101 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18570101 0.025 mg 25µg
18524044 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18570101 Supplier Novus Biologicals Supplier No. NBP1763890.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DHX58 Polyclonal antibody specifically detects DHX58 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX58
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000
Formulation PBS
Gene Accession No. EAW60798
Gene Alias D11lgp2e, D11LGP2probable ATP-dependent RNA helicase DHX58, DEXH (Asp-Glu-X-His) box polypeptide 58, EC 3.6.4.13, LGP2ortholog of mouse D11lgp2, Probable ATP-dependent helicase LGP2, Protein D11Lgp2 homolog, RIG-I-like receptor LGP2, RLR, RNA helicase LGP2
Host Species Rabbit
Immunogen Antibody was raised against a 11 amino acid synthetic peptide from near the center of human LGP2. The immunogen is located within amino acids 200 - 250 of LGP2. Amino Acid Squence: HRRSQDPFGDL
Purification Method Peptide affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 79132
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.