missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX58 Antibody (3E11F1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00€ - 578.00€
Specifications
| Antigen | DHX58 |
|---|---|
| Clone | 3E11F1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18239654
|
Novus Biologicals
NBP2-61750 |
0.1 mL |
578.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624947
|
Novus Biologicals
NBP2-61750-0.025ml |
0.025 mL |
217.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHX58 Monoclonal specifically detects DHX58 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| DHX58 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| 79132 | |
| Purified recombinant fragment of human DHX58 (AA: 479-678) expressed in E. Coli. | |
| Primary | |
| 76.6 kDa |
| 3E11F1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| D11lgp2e, D11LGP2probable ATP-dependent RNA helicase DHX58, DEXH (Asp-Glu-X-His) box polypeptide 58, EC 3.6.4.13, LGP2ortholog of mouse D11lgp2, Probable ATP-dependent helicase LGP2, Protein D11Lgp2 homolog, RIG-I-like receptor LGP2, RLR, RNA helicase LGP2 | |
| DHX58 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title