missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DGKD Monoclonal Antibody (625423)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561814
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Diacylglycerol kinases (DGKs) phosphorylate diacylglycerol (DAG) to produce phosphatidic acid. DAG and phosphatidic acid are lipids that act as second messengers in signaling cascades. DGK-alpha influences cell activation and secretion of lethal exosomes, which in turn control cell death. DGK-beta is abundant in restricted brain regions such as the caudate putamen and olfactory tubercle. DGK-gamma encodes full-length and truncated transcripts that are present in a range of human tissues, with greatest expression observed in retina. DGK-delta is most abundant in skeletal muscle. DGK-epsilon shows specificity for arachidonylcontaining diacylglycerol and is expressed predominantly in testis. DGK-theta is most abundant in the cerebellum and hippocampus. DGK-iota is present in brain and retina as a predominant transcript of more than 12 kb, including a long 3-prime untranslated region, with additional low abundance transcripts of 9.5 and 7.5 kb. DGK-eta is closely related to DGK-delta. DGK-zeta is most abundant in brain and muscle. DGKs have structural motifs that play regulatory roles, and these motifs form the basis for dividing the DGKs into five subtypes.
Specifications
| DGKD | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q16760 | |
| DGKD | |
| E. coli-derived recombinant human DGF-delta , Ile570-Thr717. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 625423 | |
| Unconjugated | |
| DGKD | |
| 130 kDa diacylglycerol kinase; AI841987; D330025K09; DAG kinase delta; DGK delta; DGKD; dgkd-2; DGKdelta; DGK-delta; Diacylglycerol kinase delta; diacylglycerol kinase, delta; diacylglycerol kinase, delta 130kDa; Diglyceride kinase; diglyceride kinase delta; EC 2.7.1.107; FLJ26930; hypothetical protein LOC513389; KIAA0145; RGD1563309 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8527 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction