missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dGK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | dGK |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16886084
|
Proteintech
16459-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16876084
|
Proteintech
16459-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mitochondrial deoxyguanosine kinase (DGUOK) is required for the phosphorylation of several deoxyribonucleosides and certain purine deoxykribonucleoside analogs widely employed as antiviral and chemotherapeutic agents. Purine deoxyribonucleoside analogs are extensively used in treatment of lymphoproliferative disorders. These compounds are administered as pro-drugs, and their efficiency is dependent on intracellular phosphorylation to the corresponding triphosphates. In mammalian cells, the phosphorylation of purine deoxyribonucleosides is mediated predomitly by 2 deoxyribonucleoside kinases: cytosolic deoxycytidine kinase (DCK) and mitochondrial deoxyguanosine kinase (DGUOK also known as DGK). DGUOK expression is ubiquitous, with highest levels in muscle, brain, liver and lymphoid tissues. Defects in DGUOK are a cause of mitochondrial DNA depletion syndrome (MDS). MDS is a clinically heterogeneous group of disorders characterized by a reduction in mitochondrial DNA (mtDNA) copy number. Primary mtDNA depletion is inherited as an autosomal recessive trait and may affect single organs, typically muscle or liver, or multiple tissues.Specifications
| dGK | |
| Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q16854, Q9QX60 | |
| 1716, 27369, 297389 | |
| dGK Fusion Protein Ag9501 | |
| Primary | |
| -20°C | |
| DGUOK |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| deoxyguanosine kinase, dGK, DGUOK | |
| DGUOK | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title