missing translation for 'onlineSavingsMsg'
Learn More

Dexketoprofen Methyl Ester, Mikromol™

Product Code. 18000660
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18000660

Brand: Mikromol MM3535.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dexketoprofen Methyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dexketoprofen Methyl Ester
CAS 81601-91-4
Analyte or Component Names Dexketoprofen Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C17 H16 O3
Certifications/Compliance ISO 17025
Synonym Dexketoprofen Methyl Ester, (S)-Methyl 2-(3-benzoylphenyl)propanoate, (alphaS)-3-Benzoyl-alpha-methylbenzeneacetic acid methyl ester, Benzeneacetic acid, 3-benzoyl-alpha-methyl-, methyl ester, (S)-, (S)-Ketoprofen methyl ester, Ketoprofen Related Compound E
InChI Formula InChI=1S/C17H16O3/c1-12(17(19)20-2)14-9-6-10-15(11-14)16(18)13-7-4-3-5-8-13/h3-12H,1-2H3/t12-/m0/s1
SMILES COC(=O)[C@@H](C)c1cccc(c1)C(=O)c2ccccc2
IUPAC Name methyl (2S)-2-(3-benzoylphenyl)propanoate
Molecular Weight (g/mol) 268.307
Quantity 100 mg
Formula Weight 268.11 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.