missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexibuprofen Methyl Ester, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Dexibuprofen Methyl Ester
Specifications
Specifications
| Chemical Name or Material | Dexibuprofen Methyl Ester |
| CAS | 81576-55-8 |
| Analyte or Component Names | Dexibuprofen Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H20 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dexibuprofen Methyl Ester, (alphaS)-alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid methyl ester, Benzeneacetic acid, alpha-methyl-4-(2-methylpropyl)-, methyl ester, (S)-, (+)-Ibuprofen methyl ester, (S)-Ibuprofen methyl ester, Methyl (S)-2-[4-(isobutylphenyl)propanoate, S-(+)-Ibuprofen methyl ester |
| InChI Formula | InChI=1S/C14H20O2/c1-10(2)9-12-5-7-13(8-6-12)11(3)14(15)16-4/h5-8,10-11H,9H2,1-4H3/t11-/m0/s1 |
| SMILES | COC(=O)[C@@H](C)c1ccc(CC(C)C)cc1 |
| IUPAC Name | methyl (2S)-2-[4-(2-methylpropyl)phenyl]propanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction