Learn More
Macherey-Nagel™ Developing Chamber
Product Code. 12760213
Change view
Click to view available options
For Use With (Equipment):
TLC Micro-Sets
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12760213 TLC Micro-Sets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12760213 Supplier Macherey-Nagel™ Supplier No. 814021
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Mason jar shaped developing chambers for TLC application. Use of small TLC plates and sheets. Dimensions: height 109 mm, diameter 63 mm, thread type GL 55, glass of hydrolytic category III, lid is made of PP with LDPE sealing disk.
Mason jar shaped developing chambers for TLC application. Use of small TLC plates and sheets. Dimensions: height 109 mm, diameter 63 mm, thread type GL 55, glass of hydrolytic category III, lid is made of PP with LDPE sealing disk.
Specifications
Quantity 4 Pk.
Type Developing Chamber
For Use With (Application) For TLC Micro-Sets
For Use With (Equipment) TLC Micro-Sets