Learn More
Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one), Mikromol™
Description
Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one), Mikromol
Specifications
Specifications
| Chemical Name or Material | Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one) |
| CAS | 224789-21-3 |
| Analyte or Component Names | Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H20 N4 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-(2-ethoxyphenyl)-5-methyl-7-propyl-, 2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one |
| InChI Formula | InChI=1S/C17H20N4O2/c1-4-8-14-18-11(3)15-17(22)19-16(20-21(14)15)12-9-6-7-10-13(12)23-5-2/h6-7,9-10H,4-5,8H2,1-3H3,(H,19,20,22) |
| Solution Type | Neat |
| SMILES | CCCc1nc(C)c2C(=O)NC(=Nn12)c3ccccc3OCC |
| Show More |