missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-3 Mouse anti-Human, Clone: DSG3/2839, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07885-100ug
This item is not returnable.
View return policy
Description
Desmoglein-3 Monoclonal specifically detects Desmoglein-3 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Desmoglein-3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
| Recombinant fragment (around aa 379-491) human Desmoglein-3 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction | |
| 1830 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Peptide Array | |
| DSG3/2839 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction