missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmethyltrimipramine Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Desmethyltrimipramine Hydrochloride |
| CAS | 10464-24-1 |
| Analyte or Component Names | Desmethyltrimipramine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H24 N2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | (2RS)-3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,2-dimethylpropan-1-amine Hydrochloride (Desmethyltrimipramine Hydrochloride), Trimipramine Maleate Imp. B (EP), Trimipramine Imp. B (EP), (2RS)-3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,2-dimethylpropan-1-amine Hydrochloride, Trimipramine Maleate Imp. B (EP) as Hydrochloride, Desmethyltrimipramine Hydrochloride, Trimipramine Maleate Impurity B as Hydrochloride, Trimipramine Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C19H24N2.ClH/c1-15(13-20-2)14-21-18-9-5-3-7-16(18)11-12-17-8-4-6-10-19(17)21;/h3-10,15,20H,11-14H2,1-2H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CNCC(C)CN1c2ccccc2CCc3ccccc13 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?