Learn More
Desmethyl Ketoprofen, TRC
Product Code. 30135225
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135225 250 mg 250mg
30136337 100 mg 100mg
17953085 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135225 Supplier Toronto Research Chemicals Supplier No. TRCD291830250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22071-22-3
Molecular Formula C15 H12 O3
Molecular Weight (g/mol) 240.25
Synonym (3-Benzoylphenyl)acetic Acid, Ketoprofen Imp. B (EP), 2-(3-Benzoylphenyl)acetic acid, RU 4462, m-Benzoylphenylacetic acid
IUPAC Name 2-(3-benzoylphenyl)acetic acid
SMILES OC(=O)Cc1cccc(c1)C(=O)c2ccccc2
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C15H12O3/c16-14(17)10-11-5-4-8-13(9-11)15(18)12-6-2-1-3-7-12/h1-9H,10H2,(H,16,17)
Formula Weight 240.0786
Percent Purity >95%
Chemical Name or Material (3-Benzoylphenyl)acetic Acid
Grade HPLC