missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DERP13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
183.00€ - 322.00€
Specifications
| Antigen | DERP13 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697507
|
Novus Biologicals
NBP3-12786-50ul |
50 μg |
183.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620267
|
Novus Biologicals
NBP3-12786-100ul |
100 μg |
322.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DERP13 Polyclonal antibody specifically detects DERP13 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| DERP13 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C7orf10, CaiB/baiF CoA-transferase family protein C7orf10, chromosome 7 open reading frame 10, Dermal papilla-derived protein 13, DERP13, DERP13FLJ11808, EC 2.-, FLJ53872, ORF19, Russel-Silver syndrome candidate | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DERP13 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 79783 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title