missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DENR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97745-100ul
This item is not returnable.
View return policy
Description
DENR Polyclonal antibody specifically detects DENR in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| DENR | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 0.5-2 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
| density-regulated protein, DRP1Protein DRP1, DRPsmooth muscle cell associated protein-3, SMAP-3, Smooth muscle cell-associated protein 3 | |
| Produced in rabbits immunized with E. coli-derived Human DENR fragment. | |
| 100 μg | |
| Signal Transduction | |
| 8562 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction