missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Delta Np73 Antibody (DNp73-1.1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11702AF488
This item is not returnable.
View return policy
Description
Delta Np73 Monoclonal antibody specifically detects Delta Np73 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Delta Np73 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| p53-related protein, p73, tumor protein p73 | |
| KLH-coupled synthetic peptide MLYVGDPARHLATC | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| DNp73-1.1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 7161 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction