missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98888-100ul
This item is not returnable.
View return policy
Description
DEK Polyclonal antibody specifically detects DEK in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| DEK | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:1500-1:50000, Immunoprecipitation 0.2-1 μL/mg of lysate | |
| D6S231EDEK oncogene (DNA binding), DEK oncogene, protein DEK | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DEK. | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 7913 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction