missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Defensin 5 Monoclonal Antibody (972207)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA560863

Product Code. 30533767

  • 441.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Defensins are a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several of the alpha defensin genes appear to be clustered on chromosome 8. The protein encoded by this gene, defensin, alpha 5, is highly expressed in the secretory granules of Paneth cells of the ileum.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Defensin 5
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q01523
DEFA5
E. coli-derived recombinant human alpha -Defensin 5, Thr64-Arg94.
100 μg
Primary
Human
Antibody
IgG1
ELISA
972207
Unconjugated
DEFA5
DEF5; DEFA5; defensin 5; defensin alpha 5; Defensin, alpha 5; defensin, alpha 5, Paneth cell-specific; defensin, alpha 5, preproprotein; defensin-5; HD5; HD-5; HD5(20-94); HD5(23-94); HD5(29-94); HD5(56-94); HD5(63-94); MGC129728
Mouse
Protein A/G
RUO
1670
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.