missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEFB4B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05559-100ul
This item is not returnable.
View return policy
Description
DEFB4B Polyclonal antibody specifically detects DEFB4B in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DEFB4B | |
| Polyclonal | |
| Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin | |
| DEFB4B defensin, beta 4B, DEFB4P | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-64 of human DEFB4A (NP_004933.1). MRVLYLLFSFLFIFLMPLPGVFGGIGDPVTCLKSGAICHPVFCPRRYKQIGTCGLPGTKCCKKP | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 100289462 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction