missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Beta-defensins (also designated BDs, or hBDs in human) are small cationic peptides with broad-spectrum antimicrobial activity against a variety of enveloped viruses, fungi and bacteria. Produced in mucosal epithelia and neutrophils of several species, beta-defensins are developmentally regulated. The family of beta-defensin proteins share a common defensin-motif that is characterized by multiple cysteine residues and a highly conserved tertiary structure. Besides playing a significant role in host immune defense, many beta-defensins also are involved in sperm maturation and capacitation. beta-defensin 121 is a 76 amino acid secreted protein that most likely contains a signal peptide sequence that requires cleavage by proteolytic enzymes in order to become biologically active. While expressed at low levels in testis, beta-defensin 121 is abundantly expressed in epididymal caput and corpus and, unlike other beta-defensins, it does not appear to be regulated by androgens.
Specifications
Specifications
| Antigen | DEFB121 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.36 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | DEFB121 |
| Gene Accession No. | Q5J5C9 |
| Gene Alias | Beta defensin 121, Beta defensin 21, DEFB 21, DEFB121, DEFB21, defensin, beta 121, ESC42 RELC |
| Gene Symbols | DEFB121 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?