missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DEFA1 (Human) Recombinant Protein
Human DEFA1 full-length ORF ( AAH27917, 1 a.a. - 94 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001667-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MRTLAILAAILLVALQAQAEPLQARADEVAAAPEQIAADIPEVVVSLAWDESLAPKHPGSRKSMDCYCRIPACIAGERRYGTCIYQGRLWAFCC
Specifications
AAH27917 | |
defensin, alpha 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DEFA1 | |
GST |
1667 | |
Wheat germ expression system | |
10 μg | |
DEF1, DEFA2, HNP-1, HP-1, MGC138393, MRS | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |